Tlačiť
Safety bunsen burner
Dodávateľ: SCHUTT LABORTECHNIK
3355202 3354202 3356202
SCUT3355202EA 1252.15 EUR
SCUT3355202 SCUT3354202 SCUT3356202
Safety bunsen burner
Ohřívače Hořáky plynové
Safety bunsen burner made of polished stainless steel and safety glass. Ideal for use in microbiological safety cabinets or in the chemical laboratory.
  • Multilingual menu navigation in most international languages
  • Easy-to-read highlighted colour display
  • Gas and air adjustment with “Click-Stop”-feature
  • Automated safety shutdown if not in use
  • Overheating protection/burner control by sensors activation 1x/2x
Schuett phoenix II eco: without display/sensor, with foot-switch
Schuett phoenix II standard: colour display, language selection, sensor and foot-More Product Information