Tlačiť
Air valves for safetycaps generation 2.0
Dodávateľ: S.C.A.T.
317010 397010 397050 397008 397100 397050-PP
SCTV317010EA 33.49 EUR
SCTV317010 SCTV397010 SCTV397050 SCTV397008 SCTV397100 SCTV397050-PP
Air valves for safetycaps generation 2.0
Ventily Vzduchové regulační ventily
These air valves can be used for analytical and preperative applications.
  • PTFE filter for clean solvents
  • Valve membrane blocks solvent vapours, opens at underpressure
  • Thread size UNF 1/4" 28G
They include a luer lock adapter which is useful for many applications.