Tlačiť
Bell Jars, with standard ground joint neck, DURAN®
Dodávateľ: DWK LIFE SCIENCES
244655907 244656106 244656903
SCOT244655907EA 306.83 EUR
SCOT244655907 SCOT244656106 SCOT244656903
Bell Jars, with standard ground joint neck, DURAN®
Nádoby Nádoby zvonové
DURAN®, borosilicate glass 3.3, clear.
  • Good chemical and physical properties
  • Very good thermal resistance
  • Quick and easy to clean
Wall thickness and geometry designed to suit vacuum applications.
The flat flange bell is used for tests with a vacuum experimental plate (vacuum chamber). The bell has a ground flange and a ground joint for the insertion of measuring equipment.
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable