Tlačiť
DURAN® Blanks for Evaporating Flasks
Dodávateľ: DWK LIFE SCIENCES
210282471 210282406 210283608 210284407 210285403 210286305
SCOT210282471EA 0 EUR
SCOT210282471 SCOT210282406 SCOT210283608 SCOT210284407 SCOT210285403 SCOT210286305
DURAN® Blanks for Evaporating Flasks
Baňky Baňky pro rotační odparky
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For glass manipulators without joint.