Tlačiť
LED ring light, VisiLED, BF S40-75
Dodávateľ: SCHOTT
400110
SCOC400110EA 914.48 EUR
SCOC400110
LED ring light, VisiLED, BF S40-75
Osvětlení/Lampy Ring Lights
The VisiLED ringlight S40-75 is the successor of the S40-55, which has been discontinued, and comes with a completely new design.
  • Slim housing; fits between two lenses for usage with objective revolver
  • 110 klux (WD 85 mm)
  • Extra homogenous illumination
  • Lifetime is typically 50000 h by actively controlling the LED‘s temperature
Designed for use with controller MC 1100 or MC 1500.
Objednávacie informácie: Controller MC 1100 or MC 1500 required.