Tlačiť
Real-Time PCR kit for detection of mycoplasma, Microsart® ATMP Mycoplasma
Dodávateľ: Sartorius
SMB95-1003
SARTSMB95-1003EA 632.49 EUR
SARTSMB95-1003
Real-Time PCR kit for detection of mycoplasma, Microsart® ATMP Mycoplasma
Mikrobiologické testovací systémy PCR Testy
The Microsart® ATMP Mycoplasma detection kit is especially designed for an easy and fast detection of mycoplasma in cell-based therapeutics like autologous chondrocyte transplants (ATMPs –advanced therapy medical products). Due to the short shelf life of these therapeutics, a fast result is needed. The PCR-technology makes it possible to reduce the time-to-result to only 3 hours.
The Microsart® ATMP Mycoplasma can be easily implemented because no specific hardware is needed. The kit can be used with any type of real-time PCR cycler that is able to detect the fluorescence dyes FAM™ and ROX™. The total testing procedure takes only 3 hours.
The kit contains all essential components in a function tested ready-to-use master mix, including the polymerase.
Certifikácie: Validated according to EP 2.6.7.