Tlačiť
Filter cartridges, Sartoclean® GF
Dodávateľ: SARTORIUS BIOPROCESS
5602504E1
SARP5602504E1EA 394.15 EUR
SARP5602504E1
Filter cartridges, Sartoclean® GF
Filtry Patrony filtrační
Sartoclean® filter cartridges are cellulose acetate (CA) membrane based prefilters. These filters are optimised for the protection of downstream equipment and Sartobran® CA final filters.
  • Ultra-low unspecific binding membrane
  • High product yields
  • Increased total throughput performance
The heterogeneous double layer is protected by a layer of glass fleece. This increased the total throughput performance of the membrane double layer and adds an adsorptive component to filtration process.