Tlačiť
Laboratory knives, PROFI series
Dodávateľ: RSG
181.086 192.031 192.033 192.036 192.053 180.532
RSGA181.086EA 35.61 EUR
RSGA181.086 233-0141 233-0142 233-0143 233-0145 RSGA180.532
Laboratory knives, PROFI series
Nože a řezačky
Chromium molybdenum vanadium steel blade, POM handle.
  • Forged, ice-tempered, hand-sharpened blades that are extremely durable and sharp
  • Dishwasher-proof
  • Different blades for various uses
These high quality forged knives are ideal for cutting foil, tubes, insulation boards and other materials. They can also be used for food applications, meat inspections and dissections.