Tlačiť
Amersham™ ECL Gold Hybridization Buffer, Cytiva
Dodávateľ: Cytiva
RPN3006
RPN3006EA 1998 EUR
RPN3006
Amersham™ ECL Gold Hybridization Buffer, Cytiva
Činidla pro elektroforézu Western Blotting Reagents
ECL Direct Nucleic Acid Labeling and Detection Systems are based on the direct labeling of DNA or RNA probes with horseradish peroxidase (HRP) in a simple 20 min chemical reaction. The resulting probe can be used without purification. Detection is achieved by generation of light via the HRP-catalyzed breakdown of luminol.
  • Direct probe labelling in a 10 minute reaction, 1 hour from hybridization to detection with ECL Direct and Hybond™ N+.
  • Eliminates handling, waste and regulatory issues associated with use of radioactivity.
  • No need to strip blots before reprobing.
  • Contains a novel rate enhancer, giving higher sensitivity with very low backgrounds on nylon membranes.
Informace o dodávce: ECL Direct is particularly useful for applications that require a blot to be reprobed several times and also for screening applications requiring rapid throughput and fast results. Each system includes the following reagents, sufficient for labeling 5 to 10 µg nucleic acid and detecting 2000 to 4000 cm ² of membrane (depending on product ordered): labeling reagent, crosslinker, control DNA, blocking agent, ECL Detection Reagents, and ECL Gold Hybridization Buffer.
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable
Aplikace Sufficient for 4000 cm² Membrane