Tlačiť
Blocking buffers
Dodávateľ: Rockland Immunochemicals
MB-070
ROCKMB-070EA 366.91 EUR
ROCKMB-070
Blocking buffers
Pufry Pufry pro elektroforézu Pufry blokovací
A blocking buffer specifically designed for western blotting using fluorochrome conjugated antibodies. Blocking buffer was prepared using ultra pure reagents dissolved in pharmaceutical grade water (WFI) and consists of a proprietary protein formulation in tris buffered saline at pH 7,6 with thimerisol added as an antimicrobial agent.