Tlačiť

Anti-IgG (H+L) Donkey Polyclonal Antibody (Texas Red®)

Dodávateľ: ROCKLAND
612-709-120
ROCK612-709-120EA 475.07 EUR
ROCK612-709-120
Anti-IgG (H+L) Donkey Polyclonal Antibody (Texas Red®)
Protilátky
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR