Tlačiť

Anti-GST Goat Polyclonal Antibody (Texas Red®)

Dodávateľ: ROCKLAND
600-109-200
ROCK600-109-200EA 1358.7 EUR
ROCK600-109-200
Anti-GST Goat Polyclonal Antibody (Texas Red®)
Protilátky
Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR