Tlačiť

Anti-IgG F(ab')2 Rabbit Polyclonal Antibody

Dodávateľ: ROCKLAND
301-4104
ROCK301-4104EA 527.94 EUR
ROCK301-4104
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR