Tlačiť

Anti-IgG (H+L) Goat Polyclonal Antibody

Dodávateľ: ROCKLAND
211-1102
ROCK211-1102EA 984.58 EUR
ROCK211-1102
Anti-IgG (H+L) Goat Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR