Tlačiť

Anti-ATP1A1 Chicken Polyclonal Antibody

Dodávateľ: ROCKLAND
200-9155
ROCK200-9155EA 2320.45 EUR
ROCK200-9155
Anti-ATP1A1 Chicken Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ATP1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Canine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR