Tlačiť
Filter discs, biplane, series 16, porosity 00, VitraPOR®
Dodávateľ: ROBU GLASFILTER
16309 16409 16609 16709 16809 16909 16509 16109 16119 16129 16209 16259
ROBU16309EA 82.92 EUR
ROBU16309 ROBU16409 ROBU16609 ROBU16709 ROBU16809 ROBU16909 ROBU16509 ROBU16109 ROBU16119 ROBU16129 ROBU16209 ROBU16259
Filter discs, biplane, series 16, porosity 00, VitraPOR®
Filtry Disky filtrační
The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for many applications.
  • Very good chemical resistance
  • Very good temperature resistance
Mase of borosilicate glass. The faces of filter discs from the series 16 are precisely ground down to tight tolerances and plane surfaces as may be required in specific installation circumstances.