Tlačiť
Hazardous Goods Bucket
Dodávateľ: RIXIUS
U7S-5101-300-K
RIXIU7S-5101-300-KEA 428.6 EUR
RIXIU7S-5101-300-K
Hazardous Goods Bucket
Kbelíky
UN bucket made of HDPE, white with shell handles, complete with lid type EH300.
  • Sturdy plastic bracket
  • Ideal bucket for transporting and storing solid hazardous substances
  • Airtight and waterproof
  • Opaque - protects your contents from light influences
  • Odourless and tasteless
  • Easy transport even when full
  • Weight: 1225 g
Suitable for food. UN - Y approval - suitable for filling goods of packaging group II and III.