Tlačiť

Kyselina boritá 20 g/l aqueous solution Kjeldahl_determination

Dodávateľ: REAGECON

Synonymá: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid

BOA25
REGEBOA25EA 179.85 EUR
REGEBOA25
Kyselina boritá 20 g/l aqueous solution Kjeldahl_determination
Kyselina boritá
Vzorec: B(OH)₃
Mw: 61.83
Teplota varu: 185
Teplota tavenia: 160
Skladovacia teplota: Ambient
Číslo MDL: MFCD00011337
Číslo CAS: 10043-35-3
EINECS: 233-139-2
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR