Tlačiť
Beakers, low form, without spout
Dodávateľ: QUARTZTECHNISCHE WERKSTATTEN
1210
QUTW1210EA 39.54 EUR
QUTW1210
Beakers, low form, without spout
Kádinky
Quartz glass.
  • High thermal shock resistance with low thermal expansion
  • High transmission from the UV to the IR spectrum range
  • High chemical resistance and purity
  • High electrical insulation properties
  • High radiation resistance
  • High temperature resistance and softening temperature
The combination of the special thermal, electrical, chemical, mechanical and optical properties in a single material makes quartz glass indispensible for high-technology applications.