Tlačiť
Disposable particle filtering respirators, P301, BizTex®
Dodávateľ: Portwest
P301WHR
PRTWP301WHREA 64.49 EUR
PRTWP301WHR
Disposable particle filtering respirators, P301, BizTex®
Polomasky a filtrační respirátory Polomasky filtrační, jednorázové Polomasky k filtraci částic
Disposable FFP3-mask consists of a PP substrate to which a laminated microporous polyethlene film is applied (PP/PE). The outer layer of fabric prevents the passage of liquid molecules inwards yet permits the vapour molecules that build up inside to pass through.
  • Adjustable nose bridge
  • Sweat absorbing nose foam (silicone/latex-free)
  • Twin elastic headband
  • Colour coded to indicate performance level
  • With easy flow exhalation valve
  • Can be used in concentrations of contaminats up to 20 times WEL
Certifikácie: EN 149:2001+A1:2009