Tlačiť
Anti-IL22R Rabbit Polyclonal Antibody
Dodávateľ: PROSCI
2497
PRSI2497EA 496.29 EUR
PRSI2497
Anti-IL22R Rabbit Polyclonal Antibody
Protilátky
IL-22 Receptor Antibody: A novel cytokine, designated IL-TIF for IL-10 related T cell-derived inducible factor and IL-22, was recently identified. The receptor for IL-22 (IL-22R, also termed CRF2-9 and IL-TIF-R1 chain) is a new member of the class II cytokine receptor family. IL-22R forms a complex with IL-10 receptor beta chain and mediates IL-22 signaling. IL-22 and its receptor activate JAK-STAT signaling pathway. IL22R is expressed in normal liver and kidney and their cell lines HepG2 and TK-10. A soluble form of IL-22 receptor, also termed IL-22 binding protein (IL-22BP) and IL-22More Product Information