Tlačiť
Calcinated petroleum coke - trace elements
Dodávateľ: LGC Standards PROMOCHEM
NIST-2719
PROONIST-2719EA 0 EUR
PROONIST-2719
Calcinated petroleum coke - trace elements
Standardy Standardy pro ropný průmysl
This reference standard is intended primarily for use in the calibration of apparatus and the evaluation of techniques employed in the analysis of calcined petroleum coke and other materials with a similar matrix.
Certifikácie: Traceable to NIST and SRM numbers.