Tlačiť
Mop frame, aluminium
Dodávateľ: PFENNIG REINIGUNGSTECHNIK
2009100 2009161
PPSA2009100EA 77.71 EUR
PPSA2009100 PPSA2009161
Mop frame, aluminium
Mopy
Aluminium mop frame with reduced weight.
  • Mechanics made of stainless steel, plate made of aluminium
  • Autoclavable
  • Contactless mop drop with MopDrop
Order Now
ŠPECIFIKÁCIA
Environmentally Preferable