Tlačiť

Isopropyl-β-D-thiogalaktopyranosid, powder, Pierce™

Dodávateľ: Thermo Fisher Scientific

Synonymá: IPTG

34060
PIER34060EA 117.36 EUR
PIER34060
Isopropyl-β-D-thiogalaktopyranosid, powder, Pierce™
Isopropyl-β-D-thiogalaktopyranosid

Thermo Scientific Pierce IPTG is isopropyl-β-D-thiogalactopyranoside for use with beta-galactosidase substrates such as X-Gal to detect expression of vectors, plasmids, or DNA fragments that contain the reporter gene.


  • X-Gal yields an intense blue precipitate to detect β-galactosidase expression
  • Can be combined with IPTG to selectively maximize the detection of β-galactosidase


The reagent X-Gal (5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside) is a chromogenic substrate for β-galactosidase that yields a blue precipitate, is soluble in DMF, and can be used to detect vectors, plasmids, or DNA fragments that contain the β-Galactosidase gene.


IPTG (Isopropyl-β-D-thiogalactopyranoside) may be used to maximize the expression of genes in expression vectors. When screening a lambda bacteriophage library with an antibody, the probability of success can be increased by preventing the expression of the fusion protein until the plaques are well established. By placing a nitrocellulose filter containing the inducer IPTG onto the plate and continuing growth at 37˚C, the expression of the potential β-galactosidase-cDNA fusion protein can be induced after 3-4 hours of plaque growth. The nitrocellulose filter is then screened with antibodies. IPTG is also used in combination with X-Gal to detect active β-galactosidase and is practical in applications using bacteriophage M13 and Igt11 vectors.


Pozor: For Research Use Only. Not for use in diagnostic procedures.

Vzorec: C₉H₁₈O₅S
Mw: 238.31
Teplota varu: 402
Číslo MDL: MFCD00063273
Číslo CAS: 367-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR