Tlačiť
Expression vectors, red firefly Luc vector for Luciferase assays, Pierce™
Dodávateľ: Thermo Fisher Scientific
16155 16156 16157
PIER16155EA 151.01 EUR
PIER16155 PIER16156 PIER16157
Expression vectors, red firefly Luc vector for Luciferase assays, Pierce™
Vektory, plazmidy a knihovny
Red Firefly Luc vectors contains a mutant form of the firefly luciferase gene that has a red-shifted emission spectrum.
  • Intracellular Red Firefly luciferase gene, optimised for high expression in mammalian systems
  • Multiple cloning site (MCS) provides versatility for transfer of regulatory elements from one plasmid to another
  • Transcription termination site (Ter), Lac operator (Lac O1), and transcriptional pause site (TPS) used to minimise background by reducing transcriptional read-through
  • Both puromycin (Pur) and ampicillin (Amp) markers for drug selection in mammalian and bacterial cells respectively
  • High-copy pUC bacterial DNA replication origin
pMCS-Red Firefly Luc is a multiple cloning site plasmid designed to accept a promoter sequence for the study of gene regulation using the intracellular red firefly luciferase reporter with excellent light intensity.
pCMV-Red Firefly Luc is a constitutive expression vector having the luciferase gene under the CMV (Cytomegalovirus) promoter. Strong (CMV) constitutive promoters for co-transfection and normalisation.
pTK-Red Firefly Luc is a constitutive expression vector having the luciferase gene under the Herpes Simplex Virus (HSV) thymidine kinase (TK) promoter. Weak (TK) constitutive promoters for co-transfection and normalisation.
Pozor: For research use only. Not for use in diagnostic procedures.