Tlačiť
Disinfectant, Phagospray DM
Dodávateľ: PHAGOGENE
60416 60414
PHAG60416EA 132.59 EUR
PHAG60416 PHAG60414
Disinfectant, Phagospray DM
Prostředky čistící Dezinfekční prostředek
Disinfectant based on aqueous alcohol for floors and large surfaces.
  • Broad spectrum disinfectant for reducing the risk of contamination
  • Contains ethanol (~23%), didecyldimethylammonium chloride, and alkylamine
  • Rapid cleaning and disinfection with a simple 'spray and wipe'
  • Grapefruit fragrance
Certifikácie: Bactericide EN 1040, EN 1276, EN 13727, EN 13697; Mycobactericide EN 14348; Fungicide EN 1275, EN 13624, EN13697; Virucide active against Rotavirus and Influenze A/H1N1 and also active against HIV and HBV.
Pozor: Use biocides safely. Always readMore Product Information