Tlačiť
Cleanroom Tables, Heavy Duty
Dodávateľ: PALBAM CLASS
WX0907 WX0907CASTORS WX0908 WX0908CASTORS WX0909 WX0909CASTORS WX0910 WX0910CASTORS WX1207 WX1207CASTORS WX1208 WX1208CASTORS WX1209 WX1209CASTORS WX1210 WX1210CASTORS WX1407 WX1407CASTORS WX1408 WX1408CASTORS WX1409 WX1409CASTORS WX1410 WX1410CASTORS WX1258 WX1607 WX0808 WX1608 WX1808 WX1508 WX1457 WX1257 WX0608 WX1558C WX0857 WX0810 WX1010 WX2008 WX1708 PCWX0610-000 PCWX0709-000 WX0709 WX0957 WX1007 WX0607 WX1108 WX0807 WX0957C WX1457C WX1158C WX1257C WX0857C WX1658 WX1758C WX2057C WX1058C WX1008 WX1258C WX1507 WX0707 WX0757 WX0708 WX2058 WX1058 WX0657C WX2007 WX1308 WX1658C WX1858C WX1358
132-0354EA 2638.89 EUR
132-0354 132-0355 132-0360 132-0361 132-0366 132-0367 132-0372 132-0373 132-0356 132-0357 132-0362 132-0363 132-0368 132-0369 132-0374 132-0375 132-0358 132-0359 132-0364 132-0365 132-0370 132-0371 132-0376 132-0377 132-1067 132-1071 132-1063 132-1072 132-1074 132-1070 132-1069 132-1066 PALBWX0608 PALBWX1558C 132-1093 132-1092 132-1094 PALBWX2008 PALBWX1708 PALBPCWX0610-000 PALBPCWX0709-000 PALBWX0709 PALBWX0957 PALBWX1007 PALBWX0607 PALBWX1108 PALBWX0807 PALBWX0957C PALBWX1457C PALBWX1158C PALBWX1257C PALBWX0857C PALBWX1658 PALBWX1758C PALBWX2057C PALBWX1058C PALBWX1008 PALBWX1258C PALBWX1507 PALBWX0707 PALBWX0757 PALBWX0708 PALBWX2058 PALBWX1058 PALBWX0657C PALBWX2007 PALBWX1308 PALBWX1658C PALBWX1858C PALBWX1358
Cleanroom Tables, Heavy Duty
Nábytek kancelářský Stoly
These cleanroom heavy duty tables are designed for more robust and heavy duty activities and are manufactured to the highest cleanroom standards. The bases are fabricated from tubing for easy cleaning and optimal airflow.
  • The tables have fixtures enabling all the accessories to be attached at time of purchase or in the future if the need arises
  • The tops are designed to reduce vibrations and are very strong holding up to 100 kg
Stainless steel 304 with electropolished finish and anodised aluminium connectors.
Informace o dodávce: Typically availableMore Product Information