Tlačiť

D(-)-Luciferin potassium salt 99,5 Endotoxin_free

Dodávateľ: OZ BIOSCIENCES

Synonymá: (S)-2-(6-Hydroxy-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid potassium salt, Beetle Luciferin potassium salt, D-Luciferin potassium, Luciferin potassium salt, Firefly luciferin potassium salt, Potassium (S)-2-(6-hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylate

LK10000
OZBILK10000EA 615.26 EUR
OZBILK10000
D(-)-Luciferin potassium salt 99,5 Endotoxin_free
D-(-)-Luciferin potassium salt

OZ Biosciences D-Luciferin K+ and Na+ salts are routinely used as Firefly’s Luciferase substrate in in vitro and in vivo bioluminescent assays. The quality and purity of the D-Luciferin are essential to obtain good and reproducible results. OZ Biosciences is offering high quality of Endotoxin-Free D-Luciferin K+ and Na+ salts.
Store at -20°C and protect from light


  • High purity > 99.5%
  • Good solubility and great sensitivity
  • Reliable in vivo reporter for bioluminescent assays
  • Endotoxin free (ideal for in vivo application)


Suitable for bioluminescent assays, whole animal imaging, ATP assays, immunoassays, pyrosequencing, luciferase fragment complementation for sequential gene analysis experiments.

Vzorec: C₁₁H₇KN₂O₃S₂
Mw: 318.41 g/mol
Číslo MDL: MFCD00044928
Číslo CAS: 115144-35-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af2d873233a094b3a909f57573bdf04fbaec5d4c&vid=812c7d41e42b1b7583f75d25f413b61aa5f82a5a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR