Tlačiť

Anti-DPP9 Mouse Monoclonal Antibody [clone: 1G9]

Dodávateľ: ORIGENE TECHNOLOGIES
CF504307
ORIGCF504307EA 950.13 EUR
ORIGCF504307
Anti-DPP9 Mouse Monoclonal Antibody [clone: 1G9]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: DPP9
Clonality: Monoclonal
Clone: 1G9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Order Now

ŠPECIFIKÁCIA

Symbol antigenu DPP9
Environmentally Preferable
Název antigenu Dipeptidylpeptidase 9
Druh protilátky Primární
Klonovalita Monoclonal
Spojení Unconjugated
Klon 1G9
Reaktivita Monkey,Human,Dog,Rat
Hostitel Mouse
Izotyp IgG1
Western blot Yes
Imunochemie Yes
Průtoková cytometrie Yes
Epitop Full length
Imunogen Full length human recombinant protein of human DPP9(NP_631898) produced in HEK293T cell.
Formulář Lyophilized
Molekulární hmotnost 96.4 kDa
Čištění purified by affinity chromatography
Křížová adsorbce No
Skladovací pufr PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
Skladovací teplota –20 °C
Transportní teplota –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR