Tlačiť
Endeavor™ Orbital Shakers
Dodávateľ: OHAUS
30770916 30770915
OHAU30770916EA 0 EUR
OHAU30770916 OHAU30770915
Endeavor™ Orbital Shakers
Třepačky Třepačky orbitální
Stable, long-term shaking with triple eccentric drive and maintenance-free brushless DC motor.
  • Adaptable to a wide variety of accessories to customize for low to mid volume applications
  • Programmable with a simple on-board menu to save time and guarantee reproducible results
  • Remote function permits PC control and data transfer
For use in a wide variety of applications including microbiology, molecular biology, cell cultures, staining/destaining, DNA studies, environmental and food sciences, chemistry, general mixing and more.
Feature a bright,
More Product Information