Tlačiť
Sample racks for chromatography vials
Dodávateľ: THERMO NATIONAL SCIENTIFIC
C4015-25
NSCAC4015-25EA 65.29 EUR
NSCAC4015-25
Sample racks for chromatography vials
Stojany Stojany na chromatografické vialky
Stackable PP or PS storage racks for 12×32 mm or 15×45 mm vials.
  • Racks feature alphanumeric indexing
  • Can be stacked for efficient storage
  • PP racks resistant to most solvents