Tlačiť

Anti-SARM1 Rabbit Polyclonal Antibody

Dodávateľ: NOVUS BIOLOGICALS
NBP2-29625 NBP2-29625SS
NOVUNBP2-29625EA 499.9 EUR
NOVUNBP2-29625 NOVUNBP2-29625SS
Anti-SARM1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SARM1 (sterile alpha and TIR motif containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR