Tlačiť

Anti-Influenza A Haemagglutinin H1N1 Rabbit Polyclonal Antibody

Dodávateľ: NOVUS BIOLOGICALS
NBP2-24955
NOVUNBP2-24955EA 495.09 EUR
NOVUNBP2-24955
Anti-Influenza A Haemagglutinin H1N1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Influenza A Haemagglutinin H1N1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR