Tlačiť

Anti-C6orf25 Rabbit Polyclonal Antibody

Dodávateľ: NOVUS BIOLOGICALS
NBP2-24647
NOVUNBP2-24647EA 495.09 EUR
NOVUNBP2-24647
Anti-C6orf25 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: C6orf25 (chromosome 6 open reading frame 25)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR