Tlačiť

Anti-GPR83 Rabbit Polyclonal Antibody

Dodávateľ: NOVUS BIOLOGICALS
NBP2-24515
NOVUNBP2-24515EA 636.49 EUR
NOVUNBP2-24515
Anti-GPR83 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GPR83 (G protein-coupled receptor 83)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR