Tlačiť

Anti-ZEB1 Mouse Monoclonal Antibody (DyLight® 650) [clone: 2A8A6]

Dodávateľ: NOVUS BIOLOGICALS
NBP2-23484C
NOVUNBP2-23484CEA 823.55 EUR
NOVUNBP2-23484C
Anti-ZEB1 Mouse Monoclonal Antibody (DyLight® 650) [clone: 2A8A6]
Protilátky
Type: Primary
Antigen: ZEB1 (zinc finger E-box binding homeobox 1)
Clonality: monoclonal
Clone: 2A8A6
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR