Tlačiť

Anti-OTUD1 Rabbit Polyclonal Antibody

Dodávateľ: NOVUS BIOLOGICALS
NBP1-90484
NOVUNBP1-90484EA 724.21 EUR
NOVUNBP1-90484
Anti-OTUD1 Rabbit Polyclonal Antibody
Protilátky
The OTUD1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to OTUD1. This antibody reacts with human. The OTUD1 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: OTUD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR