Tlačiť

Anti-GLI1 Rabbit Polyclonal Antibody (Biotin)

Dodávateľ: NOVUS BIOLOGICALS
NBP1-78259B
NOVUNBP1-78259BEA 824.75 EUR
NOVUNBP1-78259B
Anti-GLI1 Rabbit Polyclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: GLI1 (GLI family zinc finger 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR