Tlačiť

Anti-HSPA13 Rabbit Polyclonal Antibody

Dodávateľ: NOVUS BIOLOGICALS
NBP1-77455 NBP1-77455SS
NOVUNBP1-77455EA 661.73 EUR
NOVUNBP1-77455 NOVUNBP1-77455SS
Anti-HSPA13 Rabbit Polyclonal Antibody
Protilátky
Rabbit Polyclonal Hsp70 Antibody. Tested Applications: Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:400

Type: Primary
Antigen: HSP70
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR