Tlačiť

Anti-TRIM29 Goat Polyclonal Antibody

Dodávateľ: NOVUS BIOLOGICALS
NBP1-52000
NOVUNBP1-52000EA 709.39 EUR
NOVUNBP1-52000
Anti-TRIM29 Goat Polyclonal Antibody
Protilátky
The TRIM29 Antibody from Novus Biologicals is a goat polyclonal antibody to TRIM29. This antibody reacts with human. The TRIM29 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Recommended Dilutions: Immunohistochemistry (Parafin): 1 to 2 µg/ml

Type: Primary
Antigen: TRIM29
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-EKDRIKSFTTNE
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR