Tlačiť

Anti-CX3CL1 Rabbit Polyclonal Antibody

Dodávateľ: NOVUS BIOLOGICALS
NBP1-49539
NOVUNBP1-49539EA 0 EUR
NOVUNBP1-49539
Anti-CX3CL1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CX3CL1 (chemokine (C-X3-C motif) ligand 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR