Tlačiť

Anti-CDX1 Rabbit Polyclonal Antibody (DyLight® 405)

Dodávateľ: NOVUS BIOLOGICALS
NBP1-49538V
NOVUNBP1-49538VEA 823.55 EUR
NOVUNBP1-49538V
Anti-CDX1 Rabbit Polyclonal Antibody (DyLight® 405)
Protilátky
Type: Primary
Antigen: CDX1 (caudal type homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR