Tlačiť

Anti-TUBB3 Mouse Monoclonal Antibody [clone: 2E9]

Dodávateľ: NOVUS BIOLOGICALS
NBP1-42568 NBP1-42568SS
NOVUNBP1-42568EA 661.73 EUR
NOVUNBP1-42568 NOVUNBP1-42568SS
Anti-TUBB3 Mouse Monoclonal Antibody [clone: 2E9]
Protilátky
The beta-III Tubulin Antibody (2E9) from Novus Biologicals is a mouse monoclonal antibody to beta-III Tubulin. This antibody reacts with human. The beta-III Tubulin Antibody (2E9) has been validated for the following applications: Western Blot, Simple Western, Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence.

Simple Western

Type: Primary
Antigen: beta-III Tubulin
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR