Tlačiť

Anti-CDK4 Rabbit Polyclonal Antibody

Dodávateľ: NOVUS BIOLOGICALS
NBP1-31308
NOVUNBP1-31308EA 843.18 EUR
NOVUNBP1-31308
Anti-CDK4 Rabbit Polyclonal Antibody
Protilátky
The CDK4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CDK4. This antibody reacts with human, mouse, rat, bovine. The CDK4 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100 to 1:500

Type: Primary
Antigen: CDK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR