Tlačiť

Anti-Oval Cell Marker Rat Monoclonal Antibody (DyLight® 550) [clone: OC2-3C7]

Dodávateľ: NOVUS BIOLOGICALS
NBP1-18970R
NOVUNBP1-18970REA 761.87 EUR
NOVUNBP1-18970R
Anti-Oval Cell Marker Rat Monoclonal Antibody (DyLight® 550) [clone: OC2-3C7]
Protilátky
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-3C7
Conjugation: DyLight® 550
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

ŠPECIFIKÁCIA

Název antigenu Oval Cell Marker
Environmentally Preferable
Druh protilátky Primární
Klonovalita Monoclonal
Spojení DyLight® 550
Klon OC2-3C7
Reaktivita Mouse
Hostitel Rat
Izotyp IgG2a
Průtoková cytometrie Yes
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR