Tlačiť

Anti-TetR Rabbit Polyclonal Antibody (Biotin)

Dodávateľ: NOVUS BIOLOGICALS
NB600-234B
NOVUNB600-234BEA 809.93 EUR
NOVUNB600-234B
Anti-TetR Rabbit Polyclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: TetR
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR