Tlačiť

Anti-CYP46A1 Rabbit Polyclonal Antibody

Dodávateľ: NOVUS BIOLOGICALS
NB400-140 NB400-140SS
NOVUNB400-140EA 632.08 EUR
NOVUNB400-140 NOVUNB400-140SS
Anti-CYP46A1 Rabbit Polyclonal Antibody
Protilátky
The CYP46A1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CYP46A1. This antibody reacts with human, rabbit, mouse (negative). The CYP46A1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: CYP46A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rabbit, Mouse (Negative)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR