Tlačiť

Anti-SCARB1 Rabbit Polyclonal Antibody (DyLight® 650)

Dodávateľ: NOVUS BIOLOGICALS
NB400-101C
NOVUNB400-101CEA 856.8 EUR
NOVUNB400-101C
Anti-SCARB1 Rabbit Polyclonal Antibody (DyLight® 650)
Protilátky
Type: Primary
Antigen: SCARB1 (scavenger receptor class B, member 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR