Tlačiť

Anti-TUBA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: DM1A]

Dodávateľ: NOVUS BIOLOGICALS
NB100-690PE
NOVUNB100-690PEEA 741.44 EUR
NOVUNB100-690PE
Anti-TUBA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: DM1A]
Protilátky
Type: Primary
Antigen: TUBA (tubulin, alpha)
Clonality: monoclonal
Clone: DM1A
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR