Tlačiť

Anti-SIRT6 Rabbit Polyclonal Antibody (DyLight® 650)

Dodávateľ: NOVUS BIOLOGICALS
NB100-2524C
NOVUNB100-2524CEA 823.55 EUR
NOVUNB100-2524C
Anti-SIRT6 Rabbit Polyclonal Antibody (DyLight® 650)
Protilátky
Type: Primary
Antigen: SIRT6 (sirtuin 6)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR